Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145282.4 | NP_660325.4 | 5 | 614 | ||
XM_006714550.1 | XP_006714613.1 | 7 | 1004 | |||
XM_011543199.1 | XP_011541501.1 | 7 | 1000 | |||
XM_017009086.1 | XP_016864575.1 | 8 | 1134 | |||
XM_017009090.1 | XP_016864579.1 | 11 | 2246 | |||
GenBank mRNA | BC025747.1 | - | 5 | 563 | 5200129 |