Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145294.4 | NP_660337.3 | 39, 40 | 5066 | ||
XM_017023023.1 | XP_016878512.1 | 40, 41 | 5336 | |||
XM_017023024.1 | XP_016878513.1 | 22, 23 | 2949 | |||
GenBank mRNA | AK092719.1 | - | 10, 11 | 1782 | ||
AK093609.1 | - | 4, 5 | 1466 | |||
AK131510.1 | - | 22, 23 | 3372 | |||
BC065836.1 | - | 4, 5 | 978 |