Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 28 | 3036 | ||
XM_011545742.2 | XP_011544044.1 | 32 | 3318 | |||
XM_011545744.2 | XP_011544046.1 | 17 | 1918 | |||
XM_011545745.1 | XP_011544047.1 | 15 | 1686 | |||
XM_011545746.2 | XP_011544048.1 | 15 | 1789 | |||
XM_017022947.1 | XP_016878436.1 | 33 | 3391 | |||
XM_017022948.1 | XP_016878437.1 | 16 | 2490 | |||
XM_017022949.1 | XP_016878438.1 | 14 | 2761 | |||
GenBank mRNA | AK093467.1 | - | 4 | 599 | ||
AK128606.1 | - | 17 | 1904 | |||
AY360464.1 | - | 3 | 242 | |||
BC038400.1 | - | 5 | 481 | |||
BC109294.1 | - | 17 | 1811 | |||
BC109295.1 | - | 16 | 1764 |