Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012393.2 | NP_001012393.1 | 7 | 1142 | ||
NM_001319103.1 | NP_001306032.1 | 6 | 1420 | |||
NM_001319104.1 | NP_001306033.1 | 6 | 1057 | |||
NM_001319105.1 | NP_001306034.1 | 7 | 966 | |||
NM_001319106.1 | NP_001306035.1 | 6 | 1417 | |||
NM_002545.4 | NP_002536.1 | 6 | 1420 | |||
XM_006718846.2 | XP_006718909.1 | 7 | 1365 | |||
XM_011542856.2 | XP_011541158.1 | 8 | 1678 | |||
GenBank mRNA | AK289695.1 | - | 6 | 1048 | ||
AK299528.1 | - | 8 | 815 | |||
AK299908.1 | - | 7 | 929 | |||
AK314077.1 | - | 6 | 1417 | |||
AK315859.1 | - | 6 | 747 | |||
AK316273.1 | - | 6 | 844 | |||
AK316491.1 | - | 7 | 966 | |||
BC074742.2 | - | 6 | 899 | 30915458 | ||
BC074773.2 | - | 6 | 902 | 30915200 | ||
BC117254.1 | - | 6 | 898 | 40125805 | ||
BC126251.1 | - | 6 | 898 | 8991967 | ||
BC143945.1 | - | 6 | 895 | 9052463 | ||
BC143946.1 | - | 6 | 898 | 9052464 | ||
BX537377.1 | - | 7 | 990 | |||
EU562295.1 | - | 6 | 1454 | |||
EU562296.1 | - | 7 | 932 | |||
EU562297.1 | - | 7 | 869 | |||
EU562298.1 | - | 6 | 847 | |||
EU562299.1 | - | 6 | 759 | |||
EU562300.1 | - | 5 | 683 | |||
L34774.1 | - | 6 | 925 |