Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176814.4 | NP_789784.2 | 3, 4 | 888 | ||
XM_005250181.3 | XP_005250238.1 | 3, 4 | 962 | |||
XM_006715867.3 | XP_006715930.1 | 3, 4 | 967 | |||
XM_006715868.3 | XP_006715931.1 | 3, 4 | 962 | |||
XM_006715869.3 | XP_006715932.1 | 2, 3 | 871 | |||
XM_011515859.2 | XP_011514161.1 | 3, 4 | 683 | |||
GenBank mRNA | AF218032.1 | - | 3, 4 | 858 | ||
AK297946.1 | - | 1, 2 | 76 | |||
AK309674.1 | - | 3, 4 | 882 | |||
BC047641.1 | - | 3, 4 | 907 | |||
BC110623.1 | - | 3, 4 | 433 | 40036036 | ||
BC110624.1 | - | 3, 4 | 428 | 40036038 | ||
DQ655936.2 | - | 3, 4 | 279 |