Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176813.4 | NP_789783.1 | 4 | 288 | ||
XM_005249627.2 | XP_005249684.1 | 4 | 267 | |||
XM_011515152.2 | XP_011513454.1 | 4 | 280 | |||
GenBank mRNA | AY069977.1 | - | 3 | 253 | ||
AY359065.1 | - | 4 | 272 | |||
BC058284.1 | - | 4 | 293 | 4694757 |