Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079537.1 | NP_001073005.1 | 3 | 520 | ||
NM_177452.3 | NP_803235.1 | 3 | 520 | |||
XM_011536409.2 | XP_011534711.1 | 4 | 540 | |||
XM_017020964.1 | XP_016876453.1 | 3 | 493 | |||
GenBank mRNA | AK056690.1 | - | 3 | 490 | ||
AK310730.1 | - | 3 | 330 | |||
AK315900.1 | - | 3 | 448 | |||
AL833179.1 | - | 3 | 404 | |||
BC047328.1 | - | 3 | 404 | 5534151 | ||
BX161510.1 | - | 3 | 412 | |||
BX248013.1 | - | 3 | 476 |