Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 31 | 3496 | ||
XM_011545742.2 | XP_011544044.1 | 35 | 3778 | |||
XM_011545744.2 | XP_011544046.1 | 20 | 2378 | |||
XM_011545745.1 | XP_011544047.1 | 18 | 2146 | |||
XM_011545746.2 | XP_011544048.1 | 18 | 2249 | |||
XM_017022947.1 | XP_016878436.1 | 36 | 3851 | |||
XM_017022948.1 | XP_016878437.1 | 19 | 2950 | |||
XM_017022949.1 | XP_016878438.1 | 17 | 3221 | |||
GenBank mRNA | AK093467.1 | - | 7 | 1059 | ||
AK128606.1 | - | 20 | 2364 | |||
AY360464.1 | - | 6 | 702 | |||
BC038400.1 | - | 8 | 941 | |||
BC109294.1 | - | 20 | 2271 | |||
BC109295.1 | - | 19 | 2224 |