Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPUL1 SIRNA Assays ›
Gene Symbol:
HNRNPUL1
Gene Name:
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases:
E1B-AP5, E1BAP5, HNRPUL1
Chromosome Location:
Chr.19: 41262476 - 41307783 on Build GRCh38
UniGene ID:
Hs.155218
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44229
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPUL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301016.2
NP_001287945.1
14
2437
NM_001321208.1
NP_001308137.1
14
2477
NM_001321211.1
NP_001308140.1
14
2261
NM_007040.5
NP_008971.2
14
2768
NM_144732.4
NP_653333.1
14
2250
XM_005258459.3
XP_005258516.1
14
2601
XM_005258461.4
XP_005258518.1
14
2431
XM_005258463.3
XP_005258520.1
14
2422
XM_005258464.3
XP_005258521.1
14
2237
XM_005258465.4
XP_005258522.1
14
2448
XM_011526393.2
XP_011524695.1
14
2523
XM_011526394.2
XP_011524696.1
14
2278
XM_011526395.2
XP_011524697.1
14
2487
GenBank mRNA
AJ007509.1
-
14
2611
AK001229.1
-
5
1034
AK021455.1
-
3
1053
AK022863.1
-
14
2570
AK023006.1
-
14
2207
AK127057.1
-
13
2258
AK225850.1
-
14
2260
AK290729.1
-
14
2232
AK291315.1
-
15
2517
AK291762.1
-
14
2570
AK297127.1
-
14
2365
AK308156.1
-
15
2280
AL050146.1
-
7
1376
BC002564.2
-
14
2508
3139049
BC004242.1
-
2
412
BC009988.2
-
14
2414
4127873
BC014232.2
-
14
2389
4558792
BC027713.2
-
14
2335
4123077
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108477, XM_008758955, XM_008758957, XM_008758956, FQ222753.
This siRNA also targets the following transcripts in Mouse: NM_144922, BC027844, BC021506, BC078642, AK155604, and others.
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
response to virus
RNA binding
protein binding
enzyme binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline