Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204830.1 | NP_001191759.1 | 3 | 605 | ||
NM_015929.3 | NP_057013.1 | 3 | 407 | |||
NM_145197.2 | NP_660198.1 | 4 | 535 | |||
NM_145198.2 | NP_660199.1 | 4 | 523 | |||
NM_145199.2 | NP_660200.1 | 2 | 281 | |||
NR_037935.1 | 4 | 668 | ||||
NR_037936.1 | 3 | 344 | ||||
GenBank mRNA | AB017566.1 | - | 3 | 407 | ||
BC007001.1 | - | 3 | 327 | 3961227 | ||
BC009772.1 | - | 3 | 348 | 4091817 | ||
BC070145.1 | - | 3 | 385 | 4671058 | ||
BG717409.1 | - | 3 | 583 | |||
BQ422048.1 | - | 3 | 316 | |||
BQ432795.1 | - | 4 | 622 |