Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015964.3 | NP_057048.2 | 2 | 255 | ||
NM_016140.3 | NP_057224.2 | 3 | 346 | |||
GenBank mRNA | AF078846.1 | - | 2 | 192 | ||
AF132972.1 | - | 2 | 175 | |||
BC000691.2 | - | 2 | 202 | 3349849 | ||
BI754230.1 | - | 3 | 329 | |||
BI772576.1 | - | 2 | 624 | |||
BQ640401.1 | - | 2 | 255 |