Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF145732.1 | - | 1 | 162 | ||
AF148509.1 | - | 1 | 222 | |||
AK298480.1 | - | 1 | 91 | |||
AK299043.1 | - | 1 | 92 | |||
AK309808.1 | - | 1 | 91 | |||
AK315797.1 | - | 1 | 187 | |||
AY358465.1 | - | 1 | 194 | |||
BC002953.1 | - | 1 | 178 | 3533651 | ||
BM848429.1 | - | 1 | 244 | |||
CR749534.1 | - | 1 | 87 | |||
DA763005.1 | - | 1 | 207 |