Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006064.4 | NP_006055.3 | 2, 3 | 785 | ||
NM_016656.3 | NP_057740.2 | 2, 3 | 785 | |||
XM_005261973.4 | XP_005262030.1 | 2, 3 | 122 | |||
XM_017029217.1 | XP_016884706.1 | 2, 3 | 115 | |||
XM_017029218.1 | XP_016884707.1 | 2, 3 | 741 | |||
GenBank mRNA | AK289407.1 | - | 2, 3 | 718 | ||
AL831926.1 | - | 2, 3 | 724 | |||
BC034726.1 | - | 2, 3 | 716 | 3919252 | ||
BX641797.1 | - | 2, 3 | 695 | |||
X90530.1 | - | 2, 3 | 576 |