Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020208.3 | NP_064593.1 | 3 | 404 | ||
NM_022405.3 | NP_071800.1 | 3 | 404 | |||
XM_011533847.2 | XP_011532149.1 | 2 | 718 | |||
XM_011533848.2 | XP_011532150.1 | 3 | 414 | |||
GenBank mRNA | AF075260.1 | - | 3 | 282 | ||
AF125107.1 | - | 4 | 468 | |||
AJ276207.1 | - | 3 | 331 | |||
AJ276208.1 | - | 3 | 331 | |||
BC126197.1 | - | 3 | 344 | 8991913 | ||
BC136431.1 | - | 3 | 344 | 9020420 | ||
BC144087.1 | - | 3 | 344 | 9052607 | ||
DR006419.1 | - | 3 | 411 |