Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016609.4 | NP_057693.3 | 5 | 1145 | ||
NM_020372.3 | NP_065105.2 | 4 | 878 | |||
XM_005267747.4 | XP_005267804.1 | 5 | 3221 | |||
XM_005267748.4 | XP_005267805.1 | 5 | 3224 | |||
XM_017021361.1 | XP_016876850.1 | 5 | 3218 | |||
XM_017021362.1 | XP_016876851.1 | 5 | 3216 | |||
GenBank mRNA | AJ243653.1 | - | 2 | 193 | ||
BC020565.1 | - | 4 | 697 | 4156259 | ||
BC090870.1 | - | 4 | 429 | |||
BC111015.1 | - | 3 | 292 | |||
BX161416.1 | - | 5 | 1104 | |||
BX647316.1 | - | 2 | 606 |