Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025138.4 | NP_079414.3 | 6, 7 | 1323 | ||
XM_005266545.3 | XP_005266602.1 | 5, 6 | 1296 | |||
XM_011535239.2 | XP_011533541.1 | 6, 7 | 1343 | |||
GenBank mRNA | AB107354.1 | - | 6, 7 | 1323 | ||
AK074360.1 | - | 6, 7 | 1295 | |||
AK299663.1 | - | 3, 4 | 319 | |||
BC064348.1 | - | 6, 7 | 578 | 4939198 | ||
BX647471.1 | - | 6, 7 | 745 | |||
DA786460.1 | - | 3, 4 | 319 |