Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016520.2 | NP_057604.1 | 4, 5 | 342 | ||
XM_011518780.2 | XP_011517082.1 | 3, 4 | 279 | |||
GenBank mRNA | AF151054.1 | - | 4, 5 | 333 | ||
AF218421.1 | - | 4, 5 | 297 | |||
AK057004.1 | - | 4, 5 | 324 | |||
AK097577.1 | - | 1, 2 | 1104 | |||
AL137549.1 | - | 2, 3 | 128 | |||
AY632862.1 | - | 1, 2 | 33 | |||
BC007664.2 | - | 4, 5 | 295 | 3140172 | ||
BC017570.1 | - | 4, 5 | 299 | 4109447 | ||
CB128496.1 | - | 4, 5 | 342 |