Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277081.1 | NP_001264010.1 | 5 | 760 | ||
NM_001277082.1 | NP_001264011.1 | 4 | 623 | |||
NM_017509.3 | NP_059979.2 | 5 | 763 | |||
NR_102274.1 | 5 | 642 | ||||
XM_006723265.3 | XP_006723328.1 | 6 | 1564 | |||
XM_011527085.2 | XP_011525387.1 | 6 | 1560 | |||
XM_011527087.2 | XP_011525389.1 | 5 | 1313 | |||
XM_011527089.2 | XP_011525391.1 | 5 | 1434 | |||
GenBank mRNA | AF303046.1 | - | 5 | 762 | ||
AY373373.1 | - | 4 | 852 | |||
AY373374.1 | - | 4 | 715 | |||
BC069480.1 | - | 5 | 760 | 7262190 | ||
BC069507.1 | - | 5 | 763 | 7262202 | ||
BC069518.1 | - | 5 | 760 | 7262214 | ||
BC126137.1 | - | 5 | 753 | 8991853 | ||
BC144045.1 | - | 5 | 632 | |||
BC144046.1 | - | 5 | 750 | 9052566 | ||
BC144047.1 | - | 4 | 613 | |||
X75363.1 | - | 1 | 1312 |