Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171083.2 | NP_001164554.1 | 6 | 631 | ||
NM_017751.4 | NP_060221.2 | 8 | 850 | |||
NM_017951.4 | NP_060421.2 | 8 | 850 | |||
NR_033230.2 | 8 | 1843 | ||||
NR_033231.2 | 7 | 707 | ||||
NR_033232.2 | 7 | 774 | ||||
XM_011511443.2 | XP_011509745.1 | 8 | 1012 | |||
XM_011511444.2 | XP_011509746.1 | 8 | 1011 | |||
XM_011511445.2 | XP_011509747.1 | 8 | 706 | |||
XM_017004448.1 | XP_016859937.1 | 8 | 752 | |||
XM_017004449.1 | XP_016859938.1 | 8 | 752 | |||
GenBank mRNA | AB037839.1 | - | 8 | 1843 | ||
AK000304.1 | - | 8 | 687 | |||
AK000763.1 | - | 3 | 180 | |||
AK056501.1 | - | 8 | 642 | |||
AK299185.1 | - | 6 | 481 | |||
AK303519.1 | - | 7 | 611 | |||
BC040941.1 | - | 1 | 55 | |||
BC064947.1 | - | 7 | 529 | 6023304 | ||
EU367941.1 | - | 8 | 694 |
Gene Symbol: LOC150776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026922.1 | 8 | 950 | |||
GenBank mRNA | AK293811.1 | - | 8 | 687 | ||
AK300337.1 | - | 7 | 603 | |||
AK303293.1 | - | 6 | 520 | |||
AK304811.1 | - | 7 | 748 | |||
BC093080.1 | - | 8 | 673 | |||
CN337607.1 | - | 8 | 652 |