Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020135.2 | NP_064520.2 | 1 | 850 | ||
NM_130395.2 | NP_569079.1 | 1 | 850 | |||
XM_005249232.3 | XP_005249289.1 | 1 | 941 | |||
XM_017011060.1 | XP_016866549.1 | 1 | 941 | |||
GenBank mRNA | AB056152.1 | - | 1 | 851 | ||
AB209723.1 | - | 1 | 603 | |||
AF218313.1 | - | 1 | 237 | |||
AK094823.1 | - | 1 | 810 | |||
AK223593.1 | - | 1 | 784 | |||
AK315047.1 | - | 1 | 942 | |||
AK315471.1 | - | 1 | 819 | |||
BC018923.2 | - | 1 | 811 | 4109718 |