Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021233.2 | NP_067056.2 | 3 | 357 | ||
NM_058248.1 | NP_490649.1 | 1 | 107 | |||
XM_011541878.2 | XP_011540180.1 | 1 | 248 | |||
GenBank mRNA | AF274571.1 | - | 3 | 357 | ||
AF333389.1 | - | 1 | 107 | |||
BC126136.1 | - | 3 | 348 | 8991852 | ||
BC126138.1 | - | 3 | 348 | 8991854 |