Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282495.1 | NP_001269424.1 | 5 | 581 | ||
NM_001282496.1 | NP_001269425.1 | 4 | 613 | |||
NM_001282497.1 | NP_001269426.1 | 4 | 655 | |||
NM_001282499.1 | NP_001269428.1 | 4 | 655 | |||
NM_001321800.1 | NP_001308729.1 | 4 | 519 | |||
NM_001321801.1 | NP_001308730.1 | 4 | 519 | |||
NM_001321802.1 | NP_001308731.1 | 4 | 655 | |||
NM_001321803.1 | NP_001308732.1 | 4 | 519 | |||
NM_001321805.1 | NP_001308734.1 | 4 | 655 | |||
NM_024325.5 | NP_077301.4 | 4 | 655 | |||
XM_017028062.1 | XP_016883551.1 | 4 | 496 | |||
GenBank mRNA | AK096911.1 | - | 4 | 656 | ||
BC002897.2 | - | 4 | 445 | |||
BC065009.1 | - | 5 | 578 | 6166526 | ||
CD702168.1 | - | 4 | 407 | |||
DA034001.1 | - | 1 | 37 | |||
DB217060.1 | - | 4 | 519 |