Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025138.4 | NP_079414.3 | 7 | 1365 | ||
XM_005266545.3 | XP_005266602.1 | 6 | 1338 | |||
XM_011535239.2 | XP_011533541.1 | 7 | 1385 | |||
GenBank mRNA | AB107354.1 | - | 7 | 1365 | ||
AK022723.1 | - | 1 | 41 | |||
AK074360.1 | - | 7 | 1337 | |||
AK299663.1 | - | 4 | 361 | |||
BC053566.1 | - | 1 | 44 | |||
BC064348.1 | - | 7 | 620 | 4939198 | ||
BX647471.1 | - | 7 | 787 | |||
DA786460.1 | - | 4 | 361 |