Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NARF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038618.2 | NP_001033707.1 | 2, 3 | 402 | ||
NM_012336.3 | NP_036468.1 | 2, 3 | 244 | |||
NM_031968.2 | NP_114174.1 | 2, 3 | 244 | |||
XM_005256340.3 | XP_005256397.1 | 2, 3 | 410 | |||
XM_006722278.3 | XP_006722341.1 | 2, 3 | 332 | |||
XM_011523541.2 | XP_011521843.1 | 2, 3 | 334 | |||
XM_011523543.2 | XP_011521845.1 | 2, 3 | 150 | |||
XM_011523544.2 | XP_011521846.1 | 2, 3 | 341 | |||
XM_017024449.1 | XP_016879938.1 | 2, 3 | 387 | |||
XM_017024450.1 | XP_016879939.1 | 2, 3 | 387 | |||
GenBank mRNA | AF128406.1 | - | 2, 3 | 195 | ||
AK000929.1 | - | 2, 3 | 197 | |||
AK056966.1 | - | 2, 3 | 377 | |||
AK297875.1 | - | 2, 3 | 244 | |||
AK303155.1 | - | 2, 3 | 197 | |||
BC000438.2 | - | 2, 3 | 211 | 2820621 | ||
BQ228233.1 | - | 2, 3 | 377 | |||
DA695448.1 | - | 2, 3 | 415 |