Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303503.1 | NP_001290432.1 | 8 | 926 | ||
NM_001303504.1 | NP_001290433.1 | 7 | 818 | |||
NM_001303506.1 | NP_001290435.1 | 8 | 979 | |||
NM_001303507.1 | NP_001290436.1 | 7 | 868 | |||
NM_002720.2 | NP_002711.1 | 8 | 920 | |||
XM_006721061.3 | XP_006721124.1 | 7 | 805 | |||
XM_017023403.1 | XP_016878892.1 | 8 | 896 | |||
XM_017023404.1 | XP_016878893.1 | 7 | 832 | |||
GenBank mRNA | AF097996.1 | - | 8 | 842 | ||
AL519181.3 | - | 7 | 823 | |||
BC001416.2 | - | 8 | 873 | 3140324 | ||
BQ220878.1 | - | 7 | 728 | |||
BT019746.1 | - | 7 | 738 | |||
X70218.1 | - | 8 | 873 |