Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321386.1 | NP_001308315.1 | 5 | 683 | ||
NM_001321387.1 | NP_001308316.1 | 5 | 982 | |||
NM_002825.5 | NP_002816.1 | 5 | 920 | |||
XM_017012473.1 | XP_016867962.1 | 5 | 800 | |||
GenBank mRNA | AK290488.1 | - | 5 | 749 | ||
BC005916.1 | - | 5 | 908 | 4248708 | ||
BI462078.1 | - | 5 | 698 | |||
BX441769.2 | - | 5 | 685 | |||
CN413530.1 | - | 5 | 669 | |||
D90226.1 | - | 5 | 681 | |||
M57399.1 | - | 5 | 952 | |||
X52946.1 | - | 5 | 808 |