Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022133.3 | NP_071416.2 | 3 | 644 | ||
NM_152836.2 | NP_690049.1 | 2 | 398 | |||
NM_152837.2 | NP_690050.1 | 2 | 398 | |||
XM_005251282.4 | XP_005251339.1 | 2 | 438 | |||
XM_005251283.2 | XP_005251340.1 | 3 | 575 | |||
GenBank mRNA | AF305779.1 | - | 2 | 249 | ||
AK290903.1 | - | 2 | 314 | |||
AL833763.1 | - | 2 | 305 | |||
BC033630.1 | - | 2 | 306 | 5224216 | ||
BG700892.1 | - | 3 | 665 | |||
BG739930.1 | - | 2 | 312 | |||
CB962009.1 | - | 2 | 376 |