Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002830.3 | NP_002821.1 | 27 | 3636 | ||
XM_011511560.2 | XP_011509862.1 | 27 | 3541 | |||
XM_011511562.2 | XP_011509864.1 | 26 | 3475 | |||
XM_011511563.2 | XP_011509865.1 | 17 | 2999 | |||
XM_017004596.1 | XP_016860085.1 | 27 | 3562 | |||
XM_017004597.1 | XP_016860086.1 | 27 | 3068 | |||
XM_017004598.1 | XP_016860087.1 | 27 | 3541 | |||
XM_017004599.1 | XP_016860088.1 | 27 | 3047 | |||
XM_017004600.1 | XP_016860089.1 | 27 | 3026 | |||
XM_017004601.1 | XP_016860090.1 | 26 | 3517 | |||
XM_017004602.1 | XP_016860091.1 | 26 | 3496 | |||
XM_017004603.1 | XP_016860092.1 | 26 | 2981 | |||
XM_017004604.1 | XP_016860093.1 | 26 | 3451 | |||
XM_017004605.1 | XP_016860094.1 | 26 | 3347 | |||
XM_017004606.1 | XP_016860095.1 | 26 | 3472 | |||
XM_017004607.1 | XP_016860096.1 | 24 | 3233 | |||
XM_017004608.1 | XP_016860097.1 | 24 | 3190 | |||
GenBank mRNA | AK301218.1 | - | 15 | 1852 | ||
BC010674.1 | - | 27 | 3292 | 3853914 | ||
BX648614.1 | - | 21 | 3870 |