Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024852.3 | NP_079128.2 | 4 | 719 | ||
NM_177422.2 | NP_803171.1 | 3 | 598 | |||
XM_005270575.3 | XP_005270632.1 | 5 | 6173 | |||
XM_011540879.2 | XP_011539181.1 | 4 | 1601 | |||
XM_017000523.1 | XP_016856012.1 | 5 | 6189 | |||
GenBank mRNA | AK022827.1 | - | 4 | 474 | ||
AK027796.1 | - | 5 | 653 | |||
AK292870.1 | - | 4 | 685 | |||
BC025769.1 | - | 3 | 422 | |||
BC066888.1 | - | 4 | 719 |