Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUF60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136033.2 | NP_001129505.1 | 2 | 195 | ||
NM_001271096.1 | NP_001258025.1 | 2 | 173 | |||
NM_001271098.1 | NP_001258027.1 | 2 | 173 | |||
NM_001271100.1 | NP_001258029.1 | 2 | 195 | |||
NM_014281.4 | NP_055096.2 | 2 | 176 | |||
NM_078480.2 | NP_510965.1 | 2 | 176 | |||
XM_011516929.2 | XP_011515231.1 | 3 | 375 | |||
XM_011516930.2 | XP_011515232.1 | 3 | 277 | |||
XM_017013234.1 | XP_016868723.1 | 2 | 1708 | |||
XM_017013235.1 | XP_016868724.1 | 2 | 278 | |||
XM_017013236.1 | XP_016868725.1 | 2 | 1708 | |||
XM_017013237.1 | XP_016868726.1 | 3 | 437 | |||
XM_017013238.1 | XP_016868727.1 | 3 | 237 | |||
XM_017013239.1 | XP_016868728.1 | 2 | 203 | |||
XM_017013240.1 | XP_016868729.1 | 2 | 203 | |||
GenBank mRNA | AF114818.1 | - | 2 | 157 | ||
AF190744.1 | - | 2 | 85 | |||
AF217197.2 | - | 2 | 103 | |||
AK292373.1 | - | 2 | 157 | |||
AL522753.3 | - | 2 | 123 | |||
BC008875.2 | - | 2 | 142 | 4138778 | ||
BC009734.1 | - | 2 | 125 | 3877563 | ||
BC011265.1 | - | 2 | 112 | 4155549 | ||
BC035435.2 | - | 1 | 76 | |||
BG771821.1 | - | 2 | 194 | |||
BX418925.2 | - | 2 | 125 | |||
CR457143.1 | - | 2 | 93 | |||
U51586.1 | - | 1 | 90 |