Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144606.5 | NP_653207.1 | 4 | 651 | ||
NM_144997.5 | NP_659434.2 | 4 | 651 | |||
XM_011523714.2 | XP_011522016.1 | 4 | 648 | |||
XM_011523716.1 | XP_011522018.1 | 2 | 1026 | |||
XM_011523718.2 | XP_011522020.1 | 3 | 532 | |||
XM_011523719.2 | XP_011522021.1 | 4 | 648 | |||
XM_011523721.2 | XP_011522023.1 | 3 | 397 | |||
XM_017024304.1 | XP_016879793.1 | 5 | 817 | |||
XM_017024305.1 | XP_016879794.1 | 5 | 832 | |||
XM_017024306.1 | XP_016879795.1 | 5 | 875 | |||
XM_017024307.1 | XP_016879796.1 | 5 | 837 | |||
XM_017024308.1 | XP_016879797.1 | 6 | 1013 | |||
XM_017024309.1 | XP_016879798.1 | 4 | 648 | |||
GenBank mRNA | AF517523.1 | - | 4 | 602 | ||
AK126951.1 | - | 1 | 1488 | |||
AK127912.1 | - | 4 | 651 | |||
BC015687.2 | - | 4 | 602 | 4684974 | ||
CX759825.1 | - | 4 | 614 | |||
KU178888.1 | - | 1 | 147 | |||
KU178889.1 | - | 1 | 147 |