Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152896.2 | NP_690856.1 | 1, 2 | 498 | ||
NR_046386.1 | 1, 2 | 498 | ||||
XM_017014253.1 | XP_016869742.1 | 1, 2 | 501 | |||
GenBank mRNA | AB071698.1 | - | 1, 2 | 196 | ||
AF274049.1 | - | 1, 2 | 326 | |||
AK308433.1 | - | 1, 2 | 450 | |||
BC028397.2 | - | 1, 2 | 498 | 4797830 | ||
BG709183.1 | - | 1, 2 | 503 |