Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278346.1 | NP_001265275.1 | 8 | 1898 | ||
NM_153373.3 | NP_699204.1 | 8 | 1075 | |||
NR_103508.1 | 5 | 661 | ||||
NR_103509.1 | 7 | 1779 | ||||
NR_103510.1 | 10 | 1296 | ||||
XM_006714925.2 | XP_006714988.1 | 7 | 911 | |||
XM_011534669.2 | XP_011532971.1 | 8 | 1067 | |||
XM_011534670.2 | XP_011532972.1 | 7 | 1400 | |||
XM_011534671.2 | XP_011532973.1 | 8 | 1055 | |||
XM_011534672.2 | XP_011532974.1 | 8 | 853 | |||
XM_011534674.2 | XP_011532976.1 | 5 | 722 | |||
XM_011534676.2 | XP_011532978.1 | 4 | 1007 | |||
XM_017009995.1 | XP_016865484.1 | 8 | 1067 | |||
XM_017009996.1 | XP_016865485.1 | 8 | 1067 | |||
XM_017009997.1 | XP_016865486.1 | 8 | 1037 | |||
XM_017009998.1 | XP_016865487.1 | 7 | 1369 | |||
XM_017009999.1 | XP_016865488.1 | 4 | 2812 | |||
GenBank mRNA | AK023470.1 | - | 5 | 1027 | ||
AK093737.1 | - | 3 | 748 | |||
AK292061.1 | - | 8 | 1037 | |||
BC008009.1 | - | 5 | 629 | |||
BC028002.1 | - | 7 | 1730 | |||
BC037567.1 | - | 8 | 1037 | 5220434 | ||
BC067745.1 | - | 7 | 746 | |||
BC110335.1 | - | 8 | 1047 | 5753177 |