Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP57L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083535.1 | NP_001077004.1 | 5 | 906 | ||
NM_001271852.1 | NP_001258781.1 | 3 | 755 | |||
NM_001271853.1 | NP_001258782.1 | 3 | 755 | |||
NM_173830.4 | NP_776191.1 | 3 | 521 | |||
XM_005266935.4 | XP_005266992.3 | 3 | 454 | |||
XM_005266936.4 | XP_005266993.3 | 3 | 454 | |||
XM_005266937.4 | XP_005266994.3 | 3 | 454 | |||
XM_005266938.4 | XP_005266995.3 | 3 | 454 | |||
XM_006715462.3 | XP_006715525.1 | 4 | 818 | |||
XM_011535759.2 | XP_011534061.1 | 3 | 768 | |||
XM_011535763.2 | XP_011534065.2 | 3 | 454 | |||
XM_017010764.1 | XP_016866253.1 | 4 | 957 | |||
XM_017010765.1 | XP_016866254.1 | 5 | 1212 | |||
XM_017010767.1 | XP_016866256.1 | 3 | 768 | |||
XM_017010768.1 | XP_016866257.1 | 3 | 582 | |||
XM_017010769.1 | XP_016866258.1 | 3 | 521 | |||
XM_017010770.1 | XP_016866259.1 | 5 | 1212 | |||
XM_017010772.1 | XP_016866261.1 | 2 | 605 | |||
GenBank mRNA | AK289910.1 | - | 3 | 387 | ||
BC033448.2 | - | 3 | 521 | 4514050 | ||
BC064365.1 | - | 3 | 750 | 5558124 | ||
BM787514.1 | - | 5 | 563 | |||
DB204503.1 | - | 1 | 127 |