Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076678.2 | NP_001070146.1 | 4 | 1819 | ||
NM_175910.6 | NP_787106.4 | 2 | 1596 | |||
GenBank mRNA | AB209086.1 | - | 2 | 1564 | ||
AK093823.1 | - | 1 | 189 | |||
AK095616.1 | - | 1 | 348 | |||
AK127476.1 | - | 1 | 1204 | |||
AK128494.1 | - | 2 | 1582 | |||
BC112346.1 | - | 1 | 1415 | |||
BX648842.1 | - | 4 | 1809 |