Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195643.1 | NP_001182572.1 | 2 | 733 | ||
NM_176815.4 | NP_789785.1 | 2 | 620 | |||
XM_011512537.2 | XP_011510839.1 | 2 | 798 | |||
GenBank mRNA | AK122674.1 | - | 2 | 733 | ||
AL832912.1 | - | 2 | 409 | |||
BC045541.1 | - | 2 | 451 | 5262102 | ||
BC063379.1 | - | 2 | 409 | 4109721 |