Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006903.4 | NP_008834.3 | 1 | 166 | ||
NM_176866.2 | NP_789842.2 | 1 | 166 | |||
NM_176867.3 | NP_789843.2 | 1 | 166 | |||
NM_176869.2 | NP_789845.1 | 1 | 166 | |||
GenBank mRNA | AB026722.1 | - | 1 | 160 | ||
AF161473.1 | - | 1 | 110 | |||
AF217187.1 | - | 1 | 160 | |||
AK000466.1 | - | 1 | 158 | |||
AK289607.1 | - | 1 | 144 | |||
AK294097.1 | - | 1 | 84 | |||
AK297096.1 | - | 1 | 155 | |||
AK299125.1 | - | 1 | 155 | |||
AL136655.1 | - | 1 | 94 | |||
AW953776.1 | - | 1 | 136 | |||
BC022803.1 | - | 1 | 142 | |||
BC035569.1 | - | 1 | 138 | |||
BC039462.1 | - | 1 | 113 | |||
BC057219.1 | - | 1 | 159 | 4289401 | ||
BG705183.1 | - | 1 | 178 | |||
BM840112.1 | - | 1 | 164 | |||
BM848099.1 | - | 1 | 166 | |||
CN423093.1 | - | 1 | 166 |