Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other YY1AP1 SIRNA Assays ›
Gene Symbol:
YY1AP1
Gene Name:
YY1 associated protein 1
Gene Aliases:
HCCA1, HCCA2, YAP, YY1AP
Chromosome Location:
Chr.1: 155659442 - 155689032 on Build GRCh38
UniGene ID:
Hs.584927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44578
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: YY1AP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198899.1
NP_001185828.1
2
436
NM_001198900.1
NP_001185829.1
4
369
NM_001198901.1
NP_001185830.1
4
259
NM_001198902.1
NP_001185831.1
4
226
NM_001198903.1
NP_001185832.1
4
746
NM_001198904.1
NP_001185833.1
4
746
NM_001198905.1
NP_001185834.1
5
410
NM_001198906.1
NP_001185835.1
5
394
NM_018253.3
NP_060723.2
6
491
NM_139118.2
NP_620829.1
5
394
NM_139119.2
NP_620830.1
5
407
NM_139121.2
NP_620832.1
5
360
GenBank mRNA
AF203474.1
-
4
426
AF206328.1
-
5
326
AF466401.1
-
4
200
AK023976.1
-
5
362
AK297562.1
-
4
559
AK298904.1
-
5
390
AK303046.1
-
4
409
AK303386.1
-
2
414
AK309701.1
-
4
565
AK315049.1
-
5
441
AY604179.1
-
4
231
BC001655.2
-
6
421
2961648
BC001843.2
-
6
421
2961648
BC003500.1
-
5
360
BC008766.2
-
4
303
3051391
BC014906.2
-
5
339
3922542
BC044887.1
-
4
366
5276742
BG325066.1
-
4
227
CR749789.1
-
5
381
DC300265.1
-
4
259
DC346197.1
-
2
436
DC388975.1
-
5
395
DC399389.1
-
5
410
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of cell cycle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline