Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297614.2 | NP_001284543.1 | 2, 3 | 707 | ||
NM_001304479.1 | NP_001291408.1 | 2, 3 | 452 | |||
NM_020468.5 | NP_065201.1 | 2, 3 | 707 | |||
NM_153816.5 | NP_722523.1 | 2, 3 | 707 | |||
XM_005248740.3 | XP_005248797.1 | 2, 3 | 681 | |||
XM_011535977.2 | XP_011534279.1 | 1, 2 | 123 | |||
XM_011535979.1 | XP_011534281.1 | 2, 3 | 247 | |||
XM_017011095.1 | XP_016866584.1 | 1, 2 | 114 | |||
XM_017011096.1 | XP_016866585.1 | 2, 3 | 422 | |||
XM_017011097.1 | XP_016866586.1 | 2, 3 | 374 | |||
XM_017011098.1 | XP_016866587.1 | 2, 3 | 409 | |||
XM_017011099.1 | XP_016866588.1 | 2, 3 | 422 | |||
XM_017011100.1 | XP_016866589.1 | 1, 2 | 114 | |||
XM_017011101.1 | XP_016866590.1 | 1, 2 | 123 | |||
GenBank mRNA | AK000362.1 | - | 2, 3 | 396 | ||
AK095380.1 | - | 2, 3 | 780 | |||
AK295417.1 | - | 2, 3 | 181 | |||
AY044865.1 | - | 2, 3 | 248 | |||
BC005110.1 | - | 2, 3 | 436 | 3959086 | ||
BC046520.2 | - | 2, 3 | 162 | |||
BC068589.1 | - | 2, 3 | 446 | 4818366 | ||
BC095419.1 | - | 2, 3 | 292 | 5268442 | ||
BI458735.1 | - | 2, 3 | 448 | |||
DC310382.1 | - | 2, 3 | 453 |