Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139488.1 | NP_001132960.1 | 4 | 806 | ||
NM_015376.2 | NP_056191.1 | 3 | 364 | |||
NM_170672.2 | NP_733772.1 | 5 | 719 | |||
XM_005264246.3 | XP_005264303.1 | 4 | 1146 | |||
XM_011532746.2 | XP_011531048.1 | 5 | 593 | |||
XM_011532747.2 | XP_011531049.1 | 4 | 608 | |||
XM_011532748.2 | XP_011531050.2 | 4 | 454 | |||
XM_017003759.1 | XP_016859248.1 | 6 | 2069 | |||
XM_017003760.1 | XP_016859249.1 | 5 | 2541 | |||
XM_017003761.1 | XP_016859250.1 | 4 | 471 | |||
XM_017003762.1 | XP_016859251.1 | 5 | 720 | |||
XM_017003763.1 | XP_016859252.1 | 4 | 450 | |||
GenBank mRNA | AB020653.1 | - | 3 | 364 | ||
AK292983.1 | - | 5 | 630 | |||
BC027849.1 | - | 5 | 719 | 5203041 | ||
BX647990.1 | - | 4 | 414 |