Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207015.1 | NP_001193944.1 | 13 | 1999 | ||
NM_001207016.1 | NP_001193945.1 | 11 | 1789 | |||
NM_002849.3 | NP_002840.2 | 14 | 2655 | |||
NM_130846.2 | NP_570897.2 | 10 | 1921 | |||
NR_073474.1 | 11 | 2145 | ||||
XM_011538615.2 | XP_011536917.1 | 14 | 2251 | |||
GenBank mRNA | AK091647.1 | - | 5 | 894 | ||
BC110900.1 | - | 10 | 1618 | 4538615 | ||
BX571751.1 | - | 10 | 1974 | |||
CR749836.1 | - | 5 | 1054 | |||
D64053.1 | - | 14 | 2657 | |||
U42361.1 | - | 14 | 2415 | |||
U77917.1 | - | 10 | 1898 | |||
X82635.1 | - | 10 | 1878 | |||
Z79693.1 | - | 14 | 2325 |