Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177551.3 | NP_808219.1 | 1 | 577 | ||
GenBank mRNA | AK290915.1 | - | 1 | 516 | ||
AY148884.1 | - | 1 | 563 | |||
BC027965.1 | - | 1 | 563 | 5222688 | ||
BC056419.1 | - | 1 | 549 | 5747743 |