Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152896.2 | NP_690856.1 | 3 | 900 | ||
NR_046386.1 | 3 | 900 | ||||
XM_011517704.2 | XP_011516006.1 | 3 | 529 | |||
XM_017014253.1 | XP_016869742.1 | 3 | 903 | |||
XM_017014254.1 | XP_016869743.1 | 3 | 529 | |||
GenBank mRNA | AB071698.1 | - | 3 | 598 | ||
AF274049.1 | - | 3 | 728 | |||
AK308433.1 | - | 3 | 852 | |||
BC028397.2 | - | 3 | 900 | 4797830 |