Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193380.1 | NP_001180309.1 | 4 | 443 | ||
NM_153480.1 | NP_705613.1 | 4 | 443 | |||
NM_153481.1 | NP_705614.1 | 2 | 307 | |||
NM_153483.2 | NP_705616.2 | 5 | 462 | |||
NR_104198.1 | 3 | 427 | ||||
XM_006712975.1 | XP_006713038.1 | 3 | 269 | |||
XM_006712976.1 | XP_006713039.1 | 3 | 407 | |||
XM_011533361.2 | XP_011531663.1 | 4 | 359 | |||
XM_011533362.2 | XP_011531664.1 | 4 | 378 | |||
XM_011533363.2 | XP_011531665.1 | 4 | 378 | |||
XM_011533364.2 | XP_011531666.1 | 4 | 378 | |||
GenBank mRNA | AF458069.1 | - | 4 | 443 | ||
AK074238.1 | - | 4 | 423 | |||
AK096793.1 | - | 3 | 405 | |||
AK096801.1 | - | 2 | 287 | |||
AK297697.1 | - | 4 | 409 | |||
AK314476.1 | - | 4 | 428 | |||
AY358451.1 | - | 4 | 443 | |||
BC035478.1 | - | 5 | 455 | |||
BC063110.1 | - | 4 | 441 | |||
BC137077.1 | - | 5 | 405 | 9021074 | ||
BC137078.1 | - | 5 | 405 | 9021075 |