Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176877.2 | NP_795352.2 | 28, 29 | 3989 | ||
XM_005270341.3 | XP_005270398.1 | 28, 29 | 3987 | |||
XM_011540462.2 | XP_011538764.1 | 28, 29 | 3987 | |||
XM_011540463.2 | XP_011538765.1 | 28, 29 | 4175 | |||
XM_011540464.2 | XP_011538766.1 | 28, 29 | 3987 | |||
XM_011540465.2 | XP_011538767.1 | 28, 29 | 3987 | |||
XM_011540466.2 | XP_011538768.1 | 28, 29 | 3987 | |||
XM_011540467.2 | XP_011538769.1 | 28, 29 | 3987 | |||
XM_011540468.2 | XP_011538770.1 | 28, 29 | 3987 | |||
XM_016999998.1 | XP_016855487.1 | 28, 29 | 3987 | |||
XM_016999999.1 | XP_016855488.1 | 28, 29 | 3987 | |||
XM_017000000.1 | XP_016855489.1 | 23, 24 | 3233 | |||
XM_017000001.1 | XP_016855490.1 | 9, 10 | 1064 | |||
GenBank mRNA | AB044807.1 | - | 15, 16 | 2268 | ||
AF397170.1 | - | 27, 28 | 3875 | |||
AJ001306.1 | - | 28, 29 | 3989 | |||
AJ224747.1 | - | 28, 29 | 3989 | |||
AJ224748.1 | - | 28, 29 | 3989 | |||
AK130492.1 | - | 8, 9 | 951 | |||
AK293519.1 | - | 2, 3 | 305 | |||
AK304355.1 | - | 23, 24 | 3346 | |||
AK308665.1 | - | 2, 3 | 552 | |||
BC144135.1 | - | 28, 29 | 3941 |