Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005946.2 | NP_005937.2 | 3 | 278 | ||
GenBank mRNA | BC029475.1 | - | 3 | 275 | 4723818 |