Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301272.1 | NP_001288201.1 | 3 | 358 | ||
NM_005949.3 | NP_005940.1 | 3 | 316 | |||
GenBank mRNA | BC029453.1 | - | 3 | 316 | 4041046 | |
BC108722.1 | - | 1 | 1448 | |||
BM919862.1 | - | 3 | 281 | |||
BQ939879.1 | - | 3 | 261 |