Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRFAM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139320.1 | NP_647536.1 | 10 | 2410 | ||
NM_148911.1 | NP_683709.1 | 9 | 2346 | |||
XM_005254750.2 | XP_005254807.1 | 11 | 2579 | |||
XM_011522153.1 | XP_011520455.1 | 11 | 2104 | |||
XM_011522155.2 | XP_011520457.1 | 9 | 2295 | |||
GenBank mRNA | AK292984.1 | - | 10 | 2175 | ||
BQ720423.1 | - | 1 | 301 |
Gene Symbol: CHRNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000746.5 | NP_000737.1 | 10 | 2224 | ||
NM_001190455.2 | NP_001177384.1 | 10 | 2311 | |||
NR_046324.1 | 8 | 2034 | ||||
XM_011521176.2 | XP_011519478.2 | 9 | 2079 | |||
XM_011521177.2 | XP_011519479.1 | 7 | 2164 | |||
XM_017021882.1 | XP_016877371.1 | 11 | 2289 | |||
XM_017021883.1 | XP_016877372.1 | 11 | 2286 | |||
XM_017021884.1 | XP_016877373.1 | 9 | 2135 |