Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022460.3 | NP_071905.3 | 7 | 1534 | ||
XM_017004700.1 | XP_016860189.1 | 7 | 1633 | |||
GenBank mRNA | AK024311.1 | - | 6 | 946 | ||
BC050636.1 | - | 7 | 1514 | 5193161 |