Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MADCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130760.2 | NP_570116.2 | 5 | 1315 | ||
NM_130762.2 | NP_570118.1 | 4 | 1054 | |||
GenBank mRNA | AK092211.1 | - | 3 | 1550 | ||
BC136408.1 | - | 5 | 1423 | 9020396 | ||
BC137506.1 | - | 4 | 1090 | 9021506 | ||
BC137507.1 | - | 4 | 1090 | 9021507 | ||
BC142629.1 | - | 5 | 1383 | 40147551 | ||
BC144065.1 | - | 4 | 1090 | 9052585 | ||
U43628.1 | - | 5 | 1377 | |||
U82483.1 | - | 5 | 1315 |